Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Calcium alginate | 9005-35-0 | MFCD00143567 | C18H24CaO19 | 100 G
SDP

Catalog No. 5000283122
Encompass_Preferred

Calcium alginate is a pH-sensitive biopolymer hydrogel used in pharmaceutical and biomedical applications. It is stable in acidic media and soluble in basic media, enabling pH-triggered swelling and controlled release of aqueous solutions. Common uses include development of oral agent-delivery systems and biodegradable nasal packing.

  • pH-sensitive hydrogel for controlled release
  • Biopolymer matrix suitable for drug delivery
  • Biodegradable gel-transforming properties for nasal packing
  • Stable in acidic conditions and soluble in basic conditions
  • Available in multiple laboratory pack sizes
  • Store at room temperature in a dry, cool place

Catalog No. 50-002-83122 Supplier Medchemexpress LLC Supplier No. HYW133967100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Calcium alginate is a pH-sensitive biopolymer hydrogel used in pharmaceutical and biomedical applications. It is stable in acidic media and soluble in basic media, enabling pH-triggered swelling and controlled release of aqueous solutions. Common uses include development of oral agent-delivery systems and biodegradable nasal packing.

  • pH-sensitive hydrogel for controlled release
  • Biopolymer matrix suitable for drug delivery
  • Biodegradable gel-transforming properties for nasal packing
  • Stable in acidic conditions and soluble in basic conditions
  • Available in multiple laboratory pack sizes
  • Store at room temperature in a dry, cool place

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.