Learn More
Cayman Chemical Calcium Assay Kit, 192 Well
SDP
Catalog No. 502896714
Encompass_Preferred
Cayman's Calcium Assay provides a quick, reliable method for determining total calcium concentration in a variety of biological samples, as well as tissue homogenates and cell lysates. The assay utilizes an optimized variant of the well-established o-Cresolphthalein-calcium reaction in which a vivid purple complex that absorbs between 560 and 590 nm is formed in the presence of calcium.
Catalog No. 50-289-6714 Supplier Cayman Chemical Supplier No. 701220192 WELL
Cayman's Calcium Assay provides a quick, reliable method for determining total calcium concentration in a variety of biological samples, as well as tissue homogenates and cell lysates. The assay utilizes an optimized variant of the well-established o-Cresolphthalein-calcium reaction in which a vivid purple complex that absorbs between 560 and 590 nm is formed in the presence of calcium.