Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium bromide dihydrate, 99+%, for analysis, Thermo Scientific™

Catalog No. AC214880010
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC214880010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC214880010 Supplier Thermo Scientific Chemicals Supplier No. 214880010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22208-73-7
Molecular Formula Br2Ca·2H2O
Synonym calcium bromide dihydrate, br2ca.2h2o, calcium bromide 2-hydrate

Specifications

Color White
Physical Form Fine Crystalline Powder
Quantity 1 kg
Assay Percent Range 99+%
Linear Formula CaBr2·2H2O
Formula Weight 235.93
Percent Purity ≥99.0%
Grade Analytical
Chemical Name or Material Calcium bromide dihydrate, 99+%
Hazard Category Danger
Hazard Statement GHS P Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.