Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium bromide hydrate, 99.999% (metals basis)

Catalog No. AA4449722
Click to view available options
Quantity:
25 g
100 g

CAS: 71626-99-8 | Br2Ca | 199.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71626-99-8
Molecular Formula Br2Ca
Molecular Weight (g/mol) 199.89
MDL Number MFCD00149608
InChI Key WGEFECGEFUFIQW-UHFFFAOYSA-L
IUPAC Name calcium dibromide
SMILES [Ca++].[Br-].[Br-]

Specifications

Color White
Physical Form Crystalline
Quantity 100 g
Formula Weight 199.90 (Anhydrous)
Percent Purity 99.999%
Assay (metals basis)
Chemical Name or Material Calcium bromide hydrate
Hazard Category H318
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 232-164-6
RTECSNumber EV9328000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.