Learn More
Molecular Devices Molecular Devices ScanLater Western Blot Kit, Europium-Labeled Streptavidin, 20 μg
Catalog No. 5000014586
Encompass_Preferred
The ScanLater Eu-labeled streptavidin western blot evaluation kit provides europium-labeled streptavidin and optimized buffers for sensitive, time-resolved fluorescence detection of biotinylated targets on western blot membranes. It is formulated for small-scale evaluation using mini gel protocols and the ScanLater detection workflow.
  • Contains 20 μg europium-labeled streptavidin.
  • Includes 40 mL blocking buffer.
  • Includes 100 mL wash buffer.
  • Reagents sufficient for approximately 10 mini gels with the recommended protocol.
  • Designed for time-resolved fluorescence western blot detection.
Catalog No. 50-000-14586 Supplier Molecular Devices Supplier No. R8200
The ScanLater Eu-labeled streptavidin western blot evaluation kit provides europium-labeled streptavidin and optimized buffers for sensitive, time-resolved fluorescence detection of biotinylated targets on western blot membranes. It is formulated for small-scale evaluation using mini gel protocols and the ScanLater detection workflow.
  • Contains 20 μg europium-labeled streptavidin.
  • Includes 40 mL blocking buffer.
  • Includes 100 mL wash buffer.
  • Reagents sufficient for approximately 10 mini gels with the recommended protocol.
  • Designed for time-resolved fluorescence western blot detection.