Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Carbide (1/4 x 1/2in. (6.4 x 12.7mm) Granules), Fisher Chemical

Catalog No. C57500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Tin Can
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
C57-500 500 g Tin Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C57-500

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 75-20-7
Molecular Formula CaC2
MDL Number MFCD00010905
Synonym calcium carbide, 3-phenoxypropylene di acetate, glycerol phenyl ether diacetate, 3-phenoxy-1,2-propanediol diacetate, phenylglyceryl ether diacetate, 1-phenyl ether-2,3-diacetate glycerol, 1,2-propanediol, 3-phenoxy-, diacetate, 1-acetyloxy-3-phenoxypropan-2-yl acetate, quartz crystal, 3-phenoxypropane-1,2-diyl diacetate

Specifications

Melting Point 2300°C
Color Gray
Physical Form Solid
Quantity 500 g
Formula Weight 64.1
Identification Pass Test
Packaging Tin Can
Density 2.22g/cm³
Chemical Name or Material Calcium Carbide
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Water reactive. Contact with water liberates extremely flammable gases. May cause burns. Corrosive to metals. Use only under a chemical fume hood. Keep away from open flames, hot surfaces and sources of ignition. Do not allow contact with water. Protect from moisture. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:3
Flammability:3
Instability:2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.