Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Calcium Carbide
SCI_ED

Catalog No. s25219 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25219 100 g Lab Grade
S25219A 500 g Lab Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25219
Only null left
Add to Cart
Add to Cart

Reacts with water or moisture

Generate acetylene gas

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 75-20-7
Synonym calcium carbide, 3-phenoxypropylene di acetate, glycerol phenyl ether diacetate, 3-phenoxy-1,2-propanediol diacetate, phenylglyceryl ether diacetate, 1-phenyl ether-2,3-diacetate glycerol, 1,2-propanediol, 3-phenoxy-, diacetate, 1-acetyloxy-3-phenoxypropan-2-yl acetate, quartz crystal, 3-phenoxypropane-1,2-diyl diacetate

Specifications

Color Black to Gray
Quantity 100 g
Assay Percent Range >90%
Linear Formula CaC2
Formula Weight 64.08
Grade Laboratory
Packaging Lab Grade
Density 2.22g/mL
Chemical Name or Material Calcium Carbide
Safety handling Check Alert storage code Red
ShelfLife 1 year
DOTInformation DOT Class 4.3, II, : Dangerous When Wet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.