Learn More
Alkali Scientific Calcium Carbonate, 5 Kilograms
SDP
Catalog No. 5000661117
Encompass_Preferred
Calcium Carbonate, laboratory grade, is supplied in a 5 kg weight and is used in a wide range of applications, from chemical analysis to manufacturing processes. It is a key component in acid-base neutralization reactions, often used in titrations and as a pH buffer in solutions. Calcium carbonate is also widely used in the pharmaceutical industry as a dietary supplement for calcium and in antacid formulations. In addition to its chemical applications, it serves as a filler material in manufacturing and is commonly found in construction materials like cement and paint.
Catalog No. 50-006-61117 Supplier Alkali Scientific Supplier No. MCA004CP
Calcium Carbonate, laboratory grade, is supplied in a 5 kg weight and is used in a wide range of applications, from chemical analysis to manufacturing processes. It is a key component in acid-base neutralization reactions, often used in titrations and as a pH buffer in solutions. Calcium carbonate is also widely used in the pharmaceutical industry as a dietary supplement for calcium and in antacid formulations. In addition to its chemical applications, it serves as a filler material in manufacturing and is commonly found in construction materials like cement and paint.