Learn More
Strem, An Ascensus Company CAS# 471-34-1. 100g. Calcium carbonate (99.95%-Ca). MFCD00010906
SDP
Catalog No. 5090114347
Encompass
CAS# 471-34-1. 100g. Calcium carbonate (99.95%-Ca). MFCD00010906. Molecular Weight: 100.09. Molecular Formula: CaCO3. Color/form: -325 mesh white pwdr. Strem# 93-2011. http://www.strem.com/catalog/v/93-2011/
Catalog No. 50-901-14347 Supplier Strem, An Ascensus Company Supplier No. 932011100G
CAS# 471-34-1. 100g. Calcium carbonate (99.95%-Ca). MFCD00010906. Molecular Weight: 100.09. Molecular Formula: CaCO3. Color/form: -325 mesh white pwdr. Strem# 93-2011. http://www.strem.com/catalog/v/93-2011/