Learn More
Strem, An Ascensus Company CAS# 471-34-1. 10g. Calcium carbonate (99.999%-Ca) PURATREM. MFCD00010906
SDP
Catalog No. 5090110663
Encompass
CAS# 471-34-1. 10g. Calcium carbonate (99.999%-Ca) PURATREM. MFCD00010906. Molecular Weight: 100.09. Molecular Formula: CaCO3. Color/form: white pwdr. Strem# 20-0200. http://www.strem.com/catalog/v/20-0200/
Catalog No. 50-901-10663 Supplier Strem, An Ascensus Company Supplier No. 20020010G
CAS# 471-34-1. 10g. Calcium carbonate (99.999%-Ca) PURATREM. MFCD00010906. Molecular Weight: 100.09. Molecular Formula: CaCO3. Color/form: white pwdr. Strem# 20-0200. http://www.strem.com/catalog/v/20-0200/