Learn More
Sigma Aldrich Fine Chemicals Biosciences Calcium carbonate ACS reagent, chelometric standard, 99.95-100.05% dry basis | 471-34-1 | MFCD00010906 | 100g
Catalog No. 501832159
Encompass
Calcium carbonate ACS reagent, chelometric standard, 99.95-100.05% dry basis | Mol Wt: 100.09 | 471-34-1 | MFCD00010906 | 100g
Catalog No. 50-183-2159 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 398101100G
Calcium carbonate ACS reagent, chelometric standard, 99.95-100.05% dry basis | Mol Wt: 100.09 | 471-34-1 | MFCD00010906 | 100g