Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium carbonate, Certified by BAM, ≥99.5%, MilliporeSigma™ Supelco™

Catalog No. 111014237 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4237 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4237 Supplier MilliporeSigma Supelco Supplier No. 2106725G
Add to Cart
Add to Cart

Specifications

CAS 471-34-1
Density 2.93 g/mL (at 25°C)
Physical Form Neat
Linear Formula CaCO3
MDL Number MFCD00010906
Solubility Insoluble in water
Type Certified Reference Material
Quantity 25 g
Formula Weight 100.09
Percent Purity ≥99.5%
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.