Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Calcium carbonate (PT-13C) | 287389-46-2 | MFCD01075577 | 98.0% | 102.09 | 13CHCaO3 | 25mg
SDP

Catalog No. 5000270025
Encompass_Preferred

Calcium carbonate 99 8% PT-13C is the 13C labeled Calcium carbonate 99 95-100 05% PT[1]

Catalog No. 50-002-70025 Supplier Medchemexpress LLC Supplier No. HYY0287AS25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Calcium carbonate 99 8% PT-13C is the 13C labeled Calcium carbonate 99 95-100 05% PT[1]

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.