Learn More
TARGETMOL CHEMICALS INC CALCIUM CHANNEL-MODULATO 5MG
SDP
Catalog No. 502581764
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Calcium channel-modulator-1 is a calcium channel-modulator (IC500.8 uM) with specialisation to block aortic constriction. purity: 99%
Catalog No. 50-258-1764 Supplier TARGETMOL CHEMICALS INC Supplier No. T101015MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Calcium channel-modulator-1 is a calcium channel-modulator (IC500.8 uM) with specialisation to block aortic constriction. purity: 99%