Learn More
Medchemexpress LLC Calcium channel-modulator-1 | 136941-70-3 | 579.45 | 5 MG
SDP
Catalog No. 5000446618
Encompass_Preferred
Calcium channel-modulator-1 is an orally active calcium channel modulator that blocks aortic contraction with an IC50 of 0.8 μM. It can be used for the study of cardiovascular conditions.
  • Orally active calcium channel modulator
  • Blocks aortic contraction with an IC50 of 0.8 μM
  • Suitable for the study of cardiovascular conditions
  • Demonstrates a 35-37% decrease in systolic blood pressure in spontaneously hypertensive rats when administered orally (20 mg/kg)
  • Powder can be stored at -20°C for 3 years
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-46618 Supplier Medchemexpress LLC Supplier No. HYU001355MG
Calcium channel-modulator-1 is an orally active calcium channel modulator that blocks aortic contraction with an IC50 of 0.8 μM. It can be used for the study of cardiovascular conditions.
  • Orally active calcium channel modulator
  • Blocks aortic contraction with an IC50 of 0.8 μM
  • Suitable for the study of cardiovascular conditions
  • Demonstrates a 35-37% decrease in systolic blood pressure in spontaneously hypertensive rats when administered orally (20 mg/kg)
  • Powder can be stored at -20°C for 3 years
  • In solvent, store at -80°C for 6 months or -20°C for 1 month