Learn More
Medchemexpress LLC Calcium channel-modulator-1 | 136941-70-3 | 579.45 | 50 MG
SDP
Catalog No. 5000448548
Encompass_Preferred
Calcium channel-modulator-1 is an orally active calcium channel modulator that blocks aortic contraction with an IC50 of 0.8 μM. It is suitable for the study of cardiovascular conditions. Oral administration (20 mg/kg) has been shown to cause a 35-37% decrease in systolic blood pressure in spontaneously hypertensive rats.
  • Blocks aortic contraction with an IC50 of 0.8 μM
  • Useful for studying cardiovascular conditions
  • Causes a decrease in systolic blood pressure in spontaneously hypertensive rats
Catalog No. 50-004-48548 Supplier Medchemexpress LLC Supplier No. HYU0013550MG
Calcium channel-modulator-1 is an orally active calcium channel modulator that blocks aortic contraction with an IC50 of 0.8 μM. It is suitable for the study of cardiovascular conditions. Oral administration (20 mg/kg) has been shown to cause a 35-37% decrease in systolic blood pressure in spontaneously hypertensive rats.
  • Blocks aortic contraction with an IC50 of 0.8 μM
  • Useful for studying cardiovascular conditions
  • Causes a decrease in systolic blood pressure in spontaneously hypertensive rats