Learn More
AG Scientific Inc Calcium Chloride 2M Sol,5X10ML
Catalog No. 502236852
Encompass_Preferred
CAS Number: 10035-04-8; 7732-18-5Chemical Formula: CaCl2Solubility: WaterConcentration: 2MAppearance: Clear SolutionSupplied as 5 x 10 mL. Sterile filtered to 0.22 µmResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-6852 Supplier AG Scientific Inc Supplier No. C27175X10ML
CAS Number: 10035-04-8; 7732-18-5Chemical Formula: CaCl2Solubility: WaterConcentration: 2MAppearance: Clear SolutionSupplied as 5 x 10 mL. Sterile filtered to 0.22 µmResearch or further manufacturing use only, not for food or drug use.