Learn More
Alkali Scientific Calcium Chloride 2M Solution, 5 x 10 Milliliters
SDP
Catalog No. 5000659874
Encompass_Preferred
Calcium Chloride 2M Solution is an aqueous solution of calcium chloride, provided in a convenient 10mL format for laboratory use. This solution is commonly used in biochemical and molecular biology experiments, particularly in applications requiring calcium ions, such as enzyme assays, cell culture, and DNA/RNA extraction. The 2M concentration provides a controlled environment for calcium-dependent reactions, making it ideal for use in various laboratory protocols. This ready-to-use solution eliminates the need for preparation, offering a time-efficient option for researchers working with calcium in their experiments.
Catalog No. 50-006-59874 Supplier Alkali Scientific Supplier No. CAL002CP
Calcium Chloride 2M Solution is an aqueous solution of calcium chloride, provided in a convenient 10mL format for laboratory use. This solution is commonly used in biochemical and molecular biology experiments, particularly in applications requiring calcium ions, such as enzyme assays, cell culture, and DNA/RNA extraction. The 2M concentration provides a controlled environment for calcium-dependent reactions, making it ideal for use in various laboratory protocols. This ready-to-use solution eliminates the need for preparation, offering a time-efficient option for researchers working with calcium in their experiments.