Learn More
Selleck Chemical LLC Calcium chloride
Supplier: Selleck Chemical LLC S69921G
Calcium chloride (CaCl2) is an ionic compound used for calcium supplementation in conditions like hypocalcemia and hyperkalemia and as an antidote for magnesium toxicity It also functions as a co-adjuvant in hemostatic formulations and effectively treats refractory hypotension and complete heart block It enhances the activity of MoPtc1 and MoPtc2 type 2C protein phosphatases involved in the ionic stress response and MAPK signalling regulation in Magnaporthe oryzae It can also be used as a desiccant and dehydrating agent