Learn More
Selleck Chemical LLC Calcium chloride
SDP
Catalog No. 5000002509
Encompass_Preferred
Calcium chloride (CaCl2) is an ionic compound used for calcium supplementation in conditions like hypocalcemia and hyperkalemia and as an antidote for magnesium toxicity It also functions as a co-adjuvant in hemostatic formulations and effectively treats refractory hypotension and complete heart block It enhances the activity of MoPtc1 and MoPtc2 type 2C protein phosphatases involved in the ionic stress response and MAPK signalling regulation in Magnaporthe oryzae It can also be used as a desiccant and dehydrating agent
Catalog No. 50-000-02509 Supplier Selleck Chemical LLC Supplier No. S6992100MG
Calcium chloride (CaCl2) is an ionic compound used for calcium supplementation in conditions like hypocalcemia and hyperkalemia and as an antidote for magnesium toxicity It also functions as a co-adjuvant in hemostatic formulations and effectively treats refractory hypotension and complete heart block It enhances the activity of MoPtc1 and MoPtc2 type 2C protein phosphatases involved in the ionic stress response and MAPK signalling regulation in Magnaporthe oryzae It can also be used as a desiccant and dehydrating agent