Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium chloride, ACS reagent, desiccant

Catalog No. p-4813384
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC206791000 100 g
AC206795000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC206791000 Supplier Thermo Scientific Chemicals Supplier No. 206791000
Only null left
Add to Cart

CAS: 10043-52-4 | CaCl2 | 110.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
PubChem CID 5284359
ChEBI CHEBI:3312
SMILES [Cl-].[Cl-].[Ca++]

Specifications

Boiling Point >1600.0°C
Melting Point 782.0°C
Quantity 100 g
Linear Formula CaCl2
Fieser 01,104; 04,368
Merck Index 15,1661
Solubility Information Solubility in water: 740g/L (20°C). Other solubilities: freely soluble in alcohol
IUPAC Name calcium;dichloride
Formula Weight 110.99
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Calcium chloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 233-140-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.