Learn More
Alkali Scientific Calcium Chloride Anhydrous, 1 Kg
SDP
Catalog No. 5000660378
Encompass_Preferred
Calcium Chloride Anhydrous, 1 kg, is a moisture-absorbing laboratory reagent commonly used for a variety of purposes, including drying agents, chemical reactions, and as a desiccant. This high-purity anhydrous form is ideal for experiments requiring precise control over moisture content. Calcium chloride is often used in laboratory settings for drying organic solvents and as a dehydrating agent in chemical synthesis. It is also employed in various applications in the pharmaceutical, food, and environmental testing industries. This 1 kg packaging is perfect for laboratories that need a manageable quantity for frequent use, ensuring reliable results in moisture-sensitive experiments and processes.
Catalog No. 50-006-60378 Supplier Alkali Scientific Supplier No. CIU001CP
Calcium Chloride Anhydrous, 1 kg, is a moisture-absorbing laboratory reagent commonly used for a variety of purposes, including drying agents, chemical reactions, and as a desiccant. This high-purity anhydrous form is ideal for experiments requiring precise control over moisture content. Calcium chloride is often used in laboratory settings for drying organic solvents and as a dehydrating agent in chemical synthesis. It is also employed in various applications in the pharmaceutical, food, and environmental testing industries. This 1 kg packaging is perfect for laboratories that need a manageable quantity for frequent use, ensuring reliable results in moisture-sensitive experiments and processes.