Learn More
Alkali Scientific Calcium Chloride Anhydrous, 10 Kilograms
SDP
Catalog No. 5000661730
Encompass_Preferred
Calcium Chloride Anhydrous, weighing 10 kilograms, is a moisture-absorbing reagent used extensively in laboratory, industrial, and commercial applications. Its ability to absorb water makes it essential in drying processes, particularly in chemical labs where precise moisture control is required. Calcium chloride is also used in de-icing applications and as a desiccant in gas and liquid drying. In chemical synthesis, it is involved in controlling reaction environments where anhydrous conditions are necessary. The 10 kg supply ensures long-term use for experiments requiring continuous moisture control, purification, or as a drying agent for laboratory processes.
Catalog No. 50-006-61730 Supplier Alkali Scientific Supplier No. UMC007CP
Calcium Chloride Anhydrous, weighing 10 kilograms, is a moisture-absorbing reagent used extensively in laboratory, industrial, and commercial applications. Its ability to absorb water makes it essential in drying processes, particularly in chemical labs where precise moisture control is required. Calcium chloride is also used in de-icing applications and as a desiccant in gas and liquid drying. In chemical synthesis, it is involved in controlling reaction environments where anhydrous conditions are necessary. The 10 kg supply ensures long-term use for experiments requiring continuous moisture control, purification, or as a drying agent for laboratory processes.