Learn More
Alkali Scientific Calcium Chloride Anhydrous, 2.5 Kilograms
SDP
Catalog No. 5000660566
Encompass_Preferred
Calcium Chloride Anhydrous, supplied in a 2.5 kg weight, is a highly effective moisture-absorbing reagent used in a variety of laboratory and industrial processes. It is primarily used as a desiccant in chemical reactions that require anhydrous conditions. Calcium chloride is also used in the production of dry gases, water treatment, and as a component in de-icing solutions. Its ability to absorb moisture makes it indispensable in environments where precise control of humidity is critical. This 2.5 kg quantity is ideal for smaller-scale laboratory applications, ensuring an adequate supply for moisture control and drying processes.
Catalog No. 50-006-60566 Supplier Alkali Scientific Supplier No. UMC008CP
Calcium Chloride Anhydrous, supplied in a 2.5 kg weight, is a highly effective moisture-absorbing reagent used in a variety of laboratory and industrial processes. It is primarily used as a desiccant in chemical reactions that require anhydrous conditions. Calcium chloride is also used in the production of dry gases, water treatment, and as a component in de-icing solutions. Its ability to absorb moisture makes it indispensable in environments where precise control of humidity is critical. This 2.5 kg quantity is ideal for smaller-scale laboratory applications, ensuring an adequate supply for moisture control and drying processes.