Learn More
Bioworld Calcium Chloride, Anhydrous, 500 g
Catalog No. 50255668
Encompass_Preferred
Calcium Chloride, anhydrousReagent used in tissue culture media and commonly used for competent cell preparationSpecifications:Usually ships within 24 hours. This product is sold for laboratory research use only.Acidity: 8-9Appearance: White solidBoiling Point : 1935°CDensity : 2.15 g/cm3HS Code : 28272000IUPAC_Name: Calcium ChlorideMelting Point: 772 °CRTECS Number: EV9800000
Catalog No. 50-255-668 Supplier Bioworld Supplier No. 403000121
Calcium Chloride, anhydrousReagent used in tissue culture media and commonly used for competent cell preparationSpecifications:Usually ships within 24 hours. This product is sold for laboratory research use only.Acidity: 8-9Appearance: White solidBoiling Point : 1935°CDensity : 2.15 g/cm3HS Code : 28272000IUPAC_Name: Calcium ChlorideMelting Point: 772 °CRTECS Number: EV9800000