Learn More
Sigma Aldrich Fine Chemicals Biosciences Calcium chloride, 10043-52-4, MFCD00010903, 100 g
Catalog No. NC1225424
Encompass
Linear Formula: CaCl2, Molecular Weight: 110.98, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%.
Catalog No. NC1225424 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C5670100G
Linear Formula: CaCl2, Molecular Weight: 110.98, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%.