Learn More
Alkali Scientific Calcium Chloride Anhydrous, 5 Kg
SDP
Catalog No. 5000661793
Encompass_Preferred
Calcium Chloride Anhydrous is a high-purity laboratory reagent primarily used as a moisture-absorbing desiccant in various applications. This 5 kg pack is ideal for use in laboratories where dry conditions are necessary to prevent moisture-related reactions. It is widely used to absorb water in laboratory experiments, drying gases, and in the preparation of chemical reagents. Calcium chloride anhydrous is also used in chemical synthesis and as a drying agent in organic chemistry. Due to its hygroscopic nature, it is often used in air-conditioning systems, refrigeration, and for deicing purposes. This product comes in a convenient 5 kg package, making it suitable for high-demand laboratory settings.
Catalog No. 50-006-61793 Supplier Alkali Scientific Supplier No. UMC003CP
Calcium Chloride Anhydrous is a high-purity laboratory reagent primarily used as a moisture-absorbing desiccant in various applications. This 5 kg pack is ideal for use in laboratories where dry conditions are necessary to prevent moisture-related reactions. It is widely used to absorb water in laboratory experiments, drying gases, and in the preparation of chemical reagents. Calcium chloride anhydrous is also used in chemical synthesis and as a drying agent in organic chemistry. Due to its hygroscopic nature, it is often used in air-conditioning systems, refrigeration, and for deicing purposes. This product comes in a convenient 5 kg package, making it suitable for high-demand laboratory settings.