Learn More
Calcium Chloride, Anhydrous, Desiccant, 4 Mesh, Approx. 95%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
Spectrum™ Chemical Calcium Chloride, Anhydrous, Desiccant, 4 Mesh is usually a solid white powder with an extremely salty taste. It is highly hygroscopic and must be kept in tightly sealed, air tight containers. Calcium chloride has many uses but its main use is as a desiccant. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 6 x 500 g |
| Packaging | Amber Glass Bottle |
| Percent Purity | ∼95% |
| Grade | Reagent |
| Chemical Name or Material | Calcium Chloride, Anhydrous, Desiccant |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.