Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride, Anhydrous, Desiccant, 8 Mesh, Approx. 95%, Spectrum™ Chemical
SDP

Catalog No. 18601249 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-249 45 kg Fiber Drum
18-601-246 2.5 kg Poly Pail
18-601-245 6 x 500 g Amber Glass Bottle
18-601-247 4 x 2.5 kg Poly Pail
18-601-248 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-601-249 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C108045KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C1080, 10043-52-4, CaCl2

Spectrum™ Chemical Calcium Chloride, Anhydrous, Desiccant, 8 Mesh is usually a solid white powder with an extremely salty taste. It is highly hygroscopic and must be kept in tightly sealed, air tight containers. Calcium chloride has many uses but its main use is as a desiccant. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
IUPAC Name calcium dichloride
SMILES [Cl-].[Cl-].[Ca++]

Specifications

CAS Max % 100%
Quantity 45 kg
Packaging Fiber Drum
Percent Purity ∼95%
Grade Ungraded
Chemical Name or Material Calcium Chloride, Anhydrous, Desiccant
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.