Learn More
Calcium Chloride, Anhydrous (Pellets, 4-20 mesh, for Desiccators), Fisher Chemical™
Shop All Fisherbrand ProductsSpecifications
Specifications
| Physical Form | White Pellets or Beads |
| pH | 8 to 10 |
| Color | Beige |
| Boiling Point | 1600°C |
| Melting Point | 782°C |
| Density | 2.15g/mL |
| Material | Calcium Chloride |
| Quantity | 10 kg |
| Identification | Pass Test |
| Mesh Size | 4 to 20 Mesh |
| Show More |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Irritating to eyes. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention. NFPA Health:2 Flammability:0 Instability:1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.