Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Calcium chloride (CaCl2), dihydrate | 10035-04-8 | 99.9% | 147.01 | 250 MG
SDP

Catalog No. 5000347548
Encompass_Preferred

Calcium chloride dihydrate, for cell culture, 99% is a biochemical reagent. It enhances the dentinogenic differentiation potential of WMTA on cells and can be used as a biological material or organic compound for life science related research. The product enhances the dentinogenic differentiation potential of WMTA on cells and can be used for various life science related research applications.

  • Purity: 99.86%
  • CAS No.: 10035-04-8
  • Molecular weight: 147.01
  • Formula: CaCl2H4O2
  • Appearance: Solid
  • Color: White to off-white
  • Solubility: ≥ 100 mg/mL (680.23 mM) in H2O
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture).

Catalog No. 50-003-47548 Supplier Medchemexpress LLC Supplier No. 25GHY108536D
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Calcium chloride dihydrate, for cell culture, 99% is a biochemical reagent. It enhances the dentinogenic differentiation potential of WMTA on cells and can be used as a biological material or organic compound for life science related research. The product enhances the dentinogenic differentiation potential of WMTA on cells and can be used for various life science related research applications.

  • Purity: 99.86%
  • CAS No.: 10035-04-8
  • Molecular weight: 147.01
  • Formula: CaCl2H4O2
  • Appearance: Solid
  • Color: White to off-white
  • Solubility: ≥ 100 mg/mL (680.23 mM) in H2O
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.