Learn More
Sigma Aldrich Fine Chemicals Biosciences Calcium Chloride | 10043-52-4 | MFCD00010903 | 500g
Catalog No. 501881162
Encompass
Calcium Chloride | Purity: 96% | Mol Wt: 110.98 | 10043-52-4 | MFCD00010903 | 500g
Catalog No. 50-188-1162 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 383147500G
Calcium Chloride | Purity: 96% | Mol Wt: 110.98 | 10043-52-4 | MFCD00010903 | 500g