Learn More
Sigma Aldrich Fine Chemicals Biosciences Calcium chloride dihydrate | 10035-04-8 | MFCD00149613 | 500g
Catalog No. NC1772255
Encompass
Calcium chloride dihydrate | Putity: ≥99% | MW: 147.01 | 10035-04-8 | MFCD00149613 | 500g
Catalog No. NC1772255 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 223506500G
Calcium chloride dihydrate | Putity: ≥99% | MW: 147.01 | 10035-04-8 | MFCD00149613 | 500g