Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium chloride, Dihydrate, MP Biomedicals™

Catalog No. MP21946355 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21946355 500 g
ICN19481580 100 g
ICN19481590 500 g
ICN19463501 1 kg
ICN19463505 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MP21946355 Supplier MP Biomedicals, Inc Supplier No. 02194635.5
Add to Cart
Add to Cart

Calcium chloride, Dihydrate Cell Culture Reagent, ∽99%, White Powder

  • Calcium chloride is a commonly used reagent in biochemistry.
  • It plays important roles in many biological processes, including signal transduction, muscle contraction, and maintenance of cell membrane and cell wall stability.
  • Calcium chloride is used in the preparation and transformation of competent E. coli and in the transfection of eukaryotic cells with either plasmid DNA or high molecular weight genomic DNA. has been used in the stabilization and two dimensional crystallization of the NADH-ubiquinone oxidoreductase (complex I) from Escherichia coli.
  • It is also used as a dessicant, coagulant in rubber manufacturing and as an accelerator in the concrete industry.
  • Calcium Chloride is a salt of calcium and chloride.
  • It behaves as a typical ionic halide, and is solid at room temperature
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00149613
InChI Key LLSDKQJKOVVTOJ-UHFFFAOYSA-L
Synonym Calcium dichloride dihydrate

Specifications

Melting Point 176°C (decomposition)(literature)
pH 4 to 9.5 (5% aq. soln.)
Physical Form Powder
Quantity 500 g
Molecular Formula CaCl2H4O2
Solubility Information Soluble in water - 294mg/mL (clear, colorless solution) with heat liberation, ethanol, acetic acid, acetone.
Molecular Weight (g/mol) 147.01
Grade ∽99%
RTECSNumber EV9810000
storageNote1 Store at Room Temperature (15-30°C). Store Desiccated.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.