Learn More
Sigma Aldrich Fine Chemicals Biosciences Calcium chloride dihydrate puriss., meets analytical specification of Ph. Eur., USP, FCC, E509, 99-103%, <=0.0001% Al | 10035-04-8 | MFCD00149613 | 1KG
Catalog No. 501879395
Encompass
Calcium chloride dihydrate puriss., meets analytical specification of Ph. Eur., USP, FCC, E509, 99-103%, <=0.0001% Al | Purity: 99-103% | Mol Wt: 147.01 | 10035-04-8 | MFCD00149613 | 1KG
Catalog No. 50-187-9395 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 120221KG
Calcium chloride dihydrate puriss., meets analytical specification of Ph. Eur., USP, FCC, E509, 99-103%, <=0.0001% Al | Purity: 99-103% | Mol Wt: 147.01 | 10035-04-8 | MFCD00149613 | 1KG