Learn More
Alkali Scientific Calcium Chloride, Dihydrate, USP Grade, 5 Kg
SDP
Catalog No. 5000661690
Encompass_Preferred
Calcium Chloride Dihydrate, USP Grade, is a premium-grade reagent supplied in a 5 kg package for pharmaceutical and laboratory use. This product is essential in pharmaceutical formulations, particularly for treating calcium deficiencies in clinical treatments such as intravenous therapy. Calcium Chloride Dihydrate is also widely used in the preparation of buffer solutions, particularly in protein purification, enzyme assays, and molecular biology protocols. The 5 kg pack is ideal for laboratories with high throughput or pharmaceutical facilities that require large quantities of high-purity calcium chloride dihydrate for a variety of medical and research applications.
Catalog No. 50-006-61690 Supplier Alkali Scientific Supplier No. UMC006CP
Calcium Chloride Dihydrate, USP Grade, is a premium-grade reagent supplied in a 5 kg package for pharmaceutical and laboratory use. This product is essential in pharmaceutical formulations, particularly for treating calcium deficiencies in clinical treatments such as intravenous therapy. Calcium Chloride Dihydrate is also widely used in the preparation of buffer solutions, particularly in protein purification, enzyme assays, and molecular biology protocols. The 5 kg pack is ideal for laboratories with high throughput or pharmaceutical facilities that require large quantities of high-purity calcium chloride dihydrate for a variety of medical and research applications.