Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride Hexahydrate, Solstice

Catalog No. 6001924 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 kg
6 x 2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-019-24 2.5 kg
60-044-719 6 x 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-019-24 Supplier Solstice Supplier No. 120742.5KG

meets analytical specification of European Pharmacopoeia, E509, 97-103%, ≤0.0001% Al

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7774-34-7
Molecular Formula CaCl2H12O6
Molecular Weight (g/mol) 219.068
MDL Number MFCD00149616
InChI Key QHFQAJHNDKBRBO-UHFFFAOYSA-L
Synonym calcium chloride hexahydrate, antarcticite, acmc-20alis, calcium hexahydrate dichloride, calcium chloride-water 1/6, calcium chloride hydrate, puratronic, calcium chloride, hydrated, calcium chloride hydrate trace metals basis 1g
PubChem CID 6093252
IUPAC Name calcium;dichloride;hexahydrate
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Ca+2]

Specifications

Quantity 2.5 kg
UN Number NONH for all modes of transport
Formula Weight 219.08g/mol
Percent Purity 97 to 103%
Density 1.71g/mL at 25°C
Chemical Name or Material Calcium Chloride Hexahydrate, Meets analytical specification of Ph. Eur.
Hazard Statement P280-P305 + P351 + P338-P337 + P313
Precautionary Statement H319
RTECSNumber EV9830000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.