Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride, purum, Solstice

Catalog No. 6002083 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-020-83 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-020-83 Supplier Solstice Supplier No. 210741KG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

missing translation for 'validMainframeMsg'

Purum, drying agent, dehydrated, granulated,≥97.0% (KT)

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
MDL Number MFCD00010903
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
PubChem CID 5284359
ChEBI CHEBI:3312
IUPAC Name calcium;dichloride
SMILES [Cl-].[Cl-].[Ca++]

Specifications

Physical Form Granules
Quantity 1 kg
Linear Formula CaCl2
UN Number NONH for all modes of transport
Formula Weight 110.98g/mol
Percent Purity ≥97% (KT)
Grade purum
Vapor Pressure 0.01mmHg (20°C)
Chemical Name or Material Calcium Chloride
Hazard Statement P305 + P351 + P338
Precautionary Statement H319
EINECSNumber 233-140-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.