Learn More
Sigma Aldrich Fine Chemicals Biosciences Calcium chloride solution, 10043-52-4, MFCD00010903, 100mL
Catalog No. 501656810
Encompass
Empirical Formula (Hill Notation) CaCl2, Molecular Weight 110.98, BioUltra, for molecular biology, ~1 M in H2O
Catalog No. 50-165-6810 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 21115100ML
Empirical Formula (Hill Notation) CaCl2, Molecular Weight 110.98, BioUltra, for molecular biology, ~1 M in H2O