Learn More
TRIBIOSCIENCE INC Calcium Chloride (Sterile, Tissue Culture Grade), 2.5M, 100ML
SDP
Catalog No. NC2676069
Encompass
The CaCl2 Solution at a concentration of 2.5M is used for chemical transfection.
Catalog No. NC2676069 Supplier TRIBIOSCIENCE INC Supplier No. TBS5072
The CaCl2 Solution at a concentration of 2.5M is used for chemical transfection.