Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Calcium citrate tetrahydrate

Catalog No. 359734250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-973-4250G 250 g
35-973-41KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 35-973-4250G Supplier Sigma Aldrich Supplier No. 359734250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Thermal decomposition of calcium citrate tetrahydrate in dynamic air or dry nitrogen has been investigated by by tg, dta and dsc.calcium citrate tetrahydrate may be used as raw material in the preparation of nanoparticle with the nominal composition ca0.8 ba0.2 ti03 :pr3+, via sol-gel process
  • It may be employed as carbon source in a simple and effective carbonization method to prepare porous carbon without any activation process
  • It is suitable for use in the preparation of bovine serum albumin (bsa) nanoparticles

Specifications

CAS 5785-44-4
Molecular Formula C12H10Ca3O14 -+ 4H2O
Linear Formula [O2CCH2C(OH)(CO2)CH2CO2]2 Ca3 -+ 4H2O
MDL Number MFCD00150786
Quantity 250 g
Synonym Tricalcium dicitrate tetrahydrate
Molecular Weight (g/mol) 570.49
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.