Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Citrate, Tetrahydrate, Powder, USP, 97.5-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18601287 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-287 500 g Poly Bottle
18-601-288 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-601-287 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C1102500GM

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

C1102, 5785-44-4, C12H10Ca3O14·4H2O

Spectrum™ Chemical Calcium Citrate, Tetrahydrate, Powder, USP is used to treat conditions caused by low calcium levels such as bone loss (osteoporosis) and weak bones (osteomalacia/rickets). All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 5785-44-4
Molecular Formula C12H18Ca3O18
Molecular Weight (g/mol) 570.49
InChI Key LNIZKKFWMDARJV-UHFFFAOYSA-H
IUPAC Name tricalcium bis(2-hydroxypropane-1,2,3-tricarboxylate) tetrahydrate
SMILES O.O.O.O.[Ca++].[Ca++].[Ca++].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

CAS Max % 100%
Loss on Drying 10.0-13.3%
Packaging Poly Bottle
Quantity 500 g
Percent Purity 97.5-100.5%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.