Learn More
Apexbio Technology LLC Calcium D-Panthotenate,25g CAS# 137-08-6
SDP
Supplier:  Apexbio Technology LLC B599525000
Encompass_Preferred
Calcium D-Panthotenate, also named vitamin B5 Calcium, is a water-soluble vitamin for many animals [1, 2]. Pantothenic acid participates in a number of intermediary metabolic reactions as a component of coenzyme A (CoA). It has been reported that pantothenic acid exhibits radioprotective and antioxidative effects and pantothenic acid has the potential to protect against cell damage produced by reactive oxygen species [3].In vitro: Pantothenic acid (1 mM) increased biosynthesis of glutathione by boosting cell energetic in Jurkat cells [3]. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0717