Learn More
Apexbio Technology LLC Calcium D-Panthotenate 137-08-6 100g
SDP
Catalog No. 5000062866
Encompass_Preferred
Calcium D-Panthotenate (CAS 137-08-6) commonly known as calcium pantothenate or vitamin B5 is a water-soluble vitamin forming an essential structural component of coenzyme A (CoA) Through its role in CoA it participates in various metabolic processes including energy metabolism and antioxidant defense mechanisms Calcium D-Panthotenate enhances glutathione synthesis thereby exhibiting protective effects against oxidative damage and apoptosis in vitro as demonstrated in Jurkat and Ehrlich ascites tumor cells In vivo it prevents neural tube defects induced by valproic acid in mouse models and modulates intestinal enzyme activity and fatty acid synthesis
Catalog No. 50-000-62866 Supplier Apexbio Technology LLC Supplier No. B5995100000
Calcium D-Panthotenate (CAS 137-08-6) commonly known as calcium pantothenate or vitamin B5 is a water-soluble vitamin forming an essential structural component of coenzyme A (CoA) Through its role in CoA it participates in various metabolic processes including energy metabolism and antioxidant defense mechanisms Calcium D-Panthotenate enhances glutathione synthesis thereby exhibiting protective effects against oxidative damage and apoptosis in vitro as demonstrated in Jurkat and Ehrlich ascites tumor cells In vivo it prevents neural tube defects induced by valproic acid in mouse models and modulates intestinal enzyme activity and fatty acid synthesis