Learn More
Enzo Life Sciences Calcium D-(+)-pantothenate (10g). CAS: 137-08-6
SDP
Supplier:  Enzo Life Sciences ALX460003G010
Encompass_Preferred
Precursor for coenzyme A biosynthesis . Alternative name: D-(+)Pantothenic acid hemicalcium salt. Purity: ≥98% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in water. Also soluble in glycerol; slightly soluble in 100% ethanol or methanol; insoluble in acetone or ether. Long Term Storage: Ambient.
Catalog No. 50-200-9284