Learn More
Teco Diagnostics CALCIUM DIAGNOSTIC KIT
Catalog No. NC9985830
Encompass_Preferred
For the direct, colorimetric determination of calcium in human serum or urine. 2x120mL Vials of Calcium Buffer, 2x120mL Vials of Calcium Color Reagent, 1x5mL Vial of Calcium Standard
Catalog No. NC9985830 Supplier Teco Diagnostics Supplier No. C503480
For the direct, colorimetric determination of calcium in human serum or urine. 2x120mL Vials of Calcium Buffer, 2x120mL Vials of Calcium Color Reagent, 1x5mL Vial of Calcium Standard