Learn More
Medchemexpress LLC Calcium disodium edetate hydrate | 23411-34-9 | MFCD00148919 | 98.0% | 374.27 g·mol⁻¹ | C10H12CaN2O8.xH2O.2Na | 10 G
SDP
Catalog No. 5000280585
Encompass_Preferred
Calcium disodium edetate hydrate is a research-grade chelating reagent used to bind divalent metal ions in biochemical and toxicology studies. Supplied as a solid, it is commonly used for metal-chelation experiments, sample stabilization, and preparation of buffer solutions in life science research.
  • High purity (98.0%) suitable for research applications.
  • Solid form for easy weighing and accurate dosing.
  • Effective chelation of divalent metal ions such as lead and cadmium.
  • Compatible with aqueous buffers and typical laboratory solvents.
  • Store sealed, away from moisture and light; in solution: -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-80585 Supplier Medchemexpress LLC Supplier No. HYW25011910G
Calcium disodium edetate hydrate is a research-grade chelating reagent used to bind divalent metal ions in biochemical and toxicology studies. Supplied as a solid, it is commonly used for metal-chelation experiments, sample stabilization, and preparation of buffer solutions in life science research.
  • High purity (98.0%) suitable for research applications.
  • Solid form for easy weighing and accurate dosing.
  • Effective chelation of divalent metal ions such as lead and cadmium.
  • Compatible with aqueous buffers and typical laboratory solvents.
  • Store sealed, away from moisture and light; in solution: -80°C (6 months) or -20°C (1 month).