Learn More
Medchemexpress LLC Calcium disodium edetate hydrate | 23411-34-9 | MFCD00149453 | 98.0% | 374.27 g/mol | C10H12CaN2Na2O8·xH2O | 100 MG
SDP
Catalog No. 5000280858
Encompass_Preferred
Calcium disodium edetate hydrate is a chelating biochemical reagent used in life-science research to bind divalent metal ions, assist in metal analysis, and support sample preparation and detoxification studies. It is supplied as a solid with high purity and is commonly incorporated into buffer formulations and analytical workflows.
  • Binds divalent metal ions such as Ca2+ and Pb2+.
  • High purity suitable for analytical and research use.
  • Supplied as a solid in laboratory pack sizes for convenient handling.
  • Stable when stored sealed, away from moisture and light at refrigerated conditions.
  • Useful for metal chelation, sample preparation, and analytical assays.
Catalog No. 50-002-80858 Supplier Medchemexpress LLC Supplier No. HYW250119100MG
Calcium disodium edetate hydrate is a chelating biochemical reagent used in life-science research to bind divalent metal ions, assist in metal analysis, and support sample preparation and detoxification studies. It is supplied as a solid with high purity and is commonly incorporated into buffer formulations and analytical workflows.
  • Binds divalent metal ions such as Ca2+ and Pb2+.
  • High purity suitable for analytical and research use.
  • Supplied as a solid in laboratory pack sizes for convenient handling.
  • Stable when stored sealed, away from moisture and light at refrigerated conditions.
  • Useful for metal chelation, sample preparation, and analytical assays.