Learn More
Medchemexpress LLC Calcium disodium edetate hydrate | 23411-34-9 | MFCD00149453 | 98.0% | 392.28 g·mol⁻¹ | C10H14CaN2Na2O9 | 5 G
SDP
Catalog No. 5000280702
Encompass_Preferred
Calcium disodium edetate hydrate is a chelating agent used in life-science research to sequester divalent metal ions in biochemical assays, buffer formulations, and sample preparation. It is supplied as a high-purity white to off-white solid suitable for laboratory weighing and formulation.
  • High purity (98.0%) suitable for research applications.
  • Effective chelation of divalent metal ions for assay and buffer stability.
  • Available in small-scale packs convenient for laboratory use.
  • Solid form is easy to weigh and handle for solution preparation.
  • Commonly used as a reagent in analytical and preparative workflows.
Catalog No. 50-002-80702 Supplier Medchemexpress LLC Supplier No. HYW2501195G
Calcium disodium edetate hydrate is a chelating agent used in life-science research to sequester divalent metal ions in biochemical assays, buffer formulations, and sample preparation. It is supplied as a high-purity white to off-white solid suitable for laboratory weighing and formulation.
  • High purity (98.0%) suitable for research applications.
  • Effective chelation of divalent metal ions for assay and buffer stability.
  • Available in small-scale packs convenient for laboratory use.
  • Solid form is easy to weigh and handle for solution preparation.
  • Commonly used as a reagent in analytical and preparative workflows.