Learn More
Medchemexpress LLC Calcium disodium edetate hydrate | 23411-34-9 | MFCD00149454 | 98.0% | 410.3 g·mol⁻¹ | C10H16CaN2Na2O10 | 500 MG
SDP
Catalog No. 5000280574
Encompass_Preferred
Calcium disodium edetate hydrate is the calcium disodium salt of ethylenediaminetetraacetic acid (EDTA). It is a metal-chelating reagent for life-science and analytical workflows, used to sequester divalent and trivalent metal ions and to control metal availability in assays and buffer systems.
  • Purity 98.0%.
  • White to off-white solid, hydrate form.
  • Effective chelator of Ca2+, Pb2+, Cd2+, and other metal ions.
  • Useful for buffer preparation and metal sequestration in biochemical assays.
  • Available in common laboratory pack sizes including 500 MG.
  • Store sealed away from moisture and light; recommended 4°C for solids.
Catalog No. 50-002-80574 Supplier Medchemexpress LLC Supplier No. HYW250119500MG
Calcium disodium edetate hydrate is the calcium disodium salt of ethylenediaminetetraacetic acid (EDTA). It is a metal-chelating reagent for life-science and analytical workflows, used to sequester divalent and trivalent metal ions and to control metal availability in assays and buffer systems.
  • Purity 98.0%.
  • White to off-white solid, hydrate form.
  • Effective chelator of Ca2+, Pb2+, Cd2+, and other metal ions.
  • Useful for buffer preparation and metal sequestration in biochemical assays.
  • Available in common laboratory pack sizes including 500 MG.
  • Store sealed away from moisture and light; recommended 4°C for solids.